Ethyl 4-Amino-3-bromo-5-chlorobenzoate 97%

Supplier: Ambeed
Warning

A310599-100MG A310599-10G A310599-1G A310599-250MG A310599-5G
77154-358EA 5 USD
77154-358 77154-360 77154-362 77154-364 77154-366
Ethyl 4-Amino-3-bromo-5-chlorobenzoate 97%
Ethyl 4-Amino-3-bromo-5-chlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrClNO₂
MDL Number: MFCD03407826
CAS Number: 874779-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR