Ethyl 7-chloroimidazo[1,2-a]pyridine-3-carboxylate 95%

Supplier: Ambeed
Warning

A310523-100MG A310523-1G A310523-250MG A310523-5G
77154-350EA 12.36 USD
77154-350 77154-352 77154-354 77154-356
Ethyl 7-chloroimidazo[1,2-a]pyridine-3-carboxylate 95%
Ethyl 7-chloroimidazo[1,2-a]pyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂O₂
MW: 224.644 g/mol
MDL Number: MFCD22200342
CAS Number: 1296201-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR