5-Bromopyrazolo[1,5-a]pyridine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A310230-100MG A310230-1G A310230-250MG A310230-50MG
77154-338EA 235.87 USD
77154-338 77154-340 77154-342 77154-344
5-Bromopyrazolo[1,5-a]pyridine-2-carboxylic acid 95%
5-Bromopyrazolo[1,5-a]pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O₂
MW: 241.04 g/mol
MDL Number: MFCD19687637
CAS Number: 1363381-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR