5-Amino-4-bromo-3-methyl-1-phenyl-1H-pyrazole 95+%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-4-bromo-3-methyl-1-phenylpyrazole

A309259-1G A309259-250MG A309259-5G
77154-326EA 50.47 USD
77154-326 77154-328 77154-330
5-Amino-4-bromo-3-methyl-1-phenyl-1H-pyrazole 95+%
5-Amino-4-bromo-3-methyl-1-phenyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrN₃
MW: 252.12 g/mol
MDL Number: MFCD04039926
CAS Number: 69464-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR