(S)-(+)-1,13-Bis(diphenylphosphino)-7,8-dihydro-6H-dibenzo[f,h][1,5]dioxonin 98%

Supplier: Ambeed
Warning

Synonyms: (S)-1,13-Bis(diphenylphosphino)-7,8-dihydro-6H-dibenzo[f,h][1,5]dioxonine, (S)-C3-TUNEPHOS

A308624-100MG A308624-1G A308624-250MG
77154-316EA 123.6 USD
77154-316 77154-318 77154-320
(S)-(+)-1,13-Bis(diphenylphosphino)-7,8-dihydro-6H-dibenzo[f,h][1,5]dioxonin 98%
(S)-(+)-1,13-Bis(diphenylphosphino)-7,8-dihydro-6H-dibenzo[f,h][1,5]dioxonin

Precaución: for Research Use Only

Formula: C₃₉H₃₂O₂P₂
MW: 594.6200 g/mol
MDL Number: MFCD06658115
CAS Number: 486429-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR