2-Iodo-m-toluidine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Iodo-3-methylaniline

A307459-1G A307459-250MG A307459-5G
77154-300EA 94.76 USD
77154-300 77154-302 77154-304
2-Iodo-m-toluidine 95%
2-Iodo-m-toluidine

Precaución: for Research Use Only

Formula: C₇H₈IN
MW: 233.049 g/mol
MDL Number: MFCD11505640
CAS Number: 89938-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR