RO4929097 98+%

Supplier: Ambeed
Warning

Synonyms: (S)-2,2-Dimethyl-N1-(6-oxo-6,7-dihydro-5H-dibenzo[b,d]azepin-7-yl)-N3-(2,2,3,3,3-pentafluoropropyl)malonamide

A306723-10MG A306723-1MG A306723-25MG A306723-5MG A306723-2MG
77154-270EA 91.67 USD
77154-270 77154-272 77154-274 77154-276 77612-764
RO4929097 98+%
RO4929097

Precaución: for Research Use Only

Formula: C₂₂H₂₀F₅N₃O₃
MW: 469.4 g/mol
MDL Number: MFCD18384976
CAS Number: 847925-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR