2-Amino-5-fluoro-4-methoxybenzoic acid 95%

Supplier: Ambeed
Warning

A306536-100MG A306536-1G A306536-250MG A306536-5G
77154-262EA 53 USD
77154-262 77154-264 77612-762 77901-509
2-Amino-5-fluoro-4-methoxybenzoic acid 95%
2-Amino-5-fluoro-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈FNO₃
MDL Number: MFCD23105873
CAS Number: 1363380-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR