2-Chloro-N-[5-(dimethylamino)-2-nitrophenyl]-5-nitrobenzamide 95%

Supplier: Ambeed
Danger

A306074-1G A306074-250MG
77154-258EA 248.23 USD
77154-258 77154-260
2-Chloro-N-[5-(dimethylamino)-2-nitrophenyl]-5-nitrobenzamide 95%
2-Chloro-N-[5-(dimethylamino)-2-nitrophenyl]-5-nitrobenzamide

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClN₄O₅
MW: 364.741 g/mol
MDL Number: MFCD27918643
CAS Number: 1079264-82-6
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR