1,2-Bis(2-chloro-5-methoxypyrimidin-4-yl)hydrazine 95%

Supplier: Ambeed
Warning

A304568-100MG A304568-1G A304568-250MG
77154-250EA 72 USD
77154-250 77154-252 77154-254
1,2-Bis(2-chloro-5-methoxypyrimidin-4-yl)hydrazine 95%
1,2-Bis(2-chloro-5-methoxypyrimidin-4-yl)hydrazine

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂N₆O₂
MW: 317.13 g/mol
MDL Number: MFCD29044964
CAS Number: 1956386-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR