Methyl 5-chloro-2-(methylamino)benzoate 95%

Supplier: Ambeed
Warning

A304375-1G A304375-250MG
77154-246EA 109.18 USD
77154-246 77154-248
Methyl 5-chloro-2-(methylamino)benzoate 95%
Methyl 5-chloro-2-(methylamino)benzoate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.63 g/mol
MDL Number: MFCD07439818
CAS Number: 55150-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR