Diethylene glycol-2-bromoethyl methyl ether 94%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3,6,9-trioxadecane, 2-[2-(2-Methoxyethoxy)ethoxy]ethyl bromide, Diethylene Glycol 2-Bromoethyl Methyl Ether, mPEG3-bromide, Methyl-PEG3-bromide

A304298-1G A304298-5G A304298-25G
77154-242EA 26.78 USD
77154-242 77154-244 77571-848
Diethylene glycol-2-bromoethyl methyl ether 94%
Diethylene glycol-2-bromoethyl methyl ether

Precaución: for Research Use Only

Formula: C₇H₁₅BrO₃
MW: 227.10 g/mol
MDL Number: MFCD00956116
CAS Number: 72593-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR