MPEP hydrochloride 98%

Supplier: Ambeed
Warning

A303368-25MG A303368-50MG A303368-1MG A303368-10MG
77154-234EA 18 USD
77154-234 77154-236 77612-750 77685-869
MPEP hydrochloride 98%
MPEP hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₁₁N.HCl
MDL Number: MFCD02262119
CAS Number: 219911-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR