1-Ethyl-3-methylimidazolium hydrogen sulfate 98%

Supplier: Ambeed
Danger

Synonyms: 1-Ethyl-3-methylimidazolium Hydrogen sulphate

A302985-10G A302985-25G A302985-5G
77154-228EA 20.6 USD
77154-228 77154-230 77154-232
1-Ethyl-3-methylimidazolium hydrogen sulfate 98%
1-Ethyl-3-methylimidazolium hydrogen sulphate

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O₄S
MW: 208.24 g/mol
MDL Number: MFCD06798195
CAS Number: 412009-61-1
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR