N-(2-Chloro-5-Nitrophenyl)Benzamide 95%

Supplier: Ambeed
Warning

A302709-10G A302709-1G A302709-250MG A302709-5G
77154-210EA 10.3 USD
77154-210 77154-212 77154-214 77154-216 77154-218
N-(2-Chloro-5-Nitrophenyl)Benzamide 95%
N-(2-Chloro-5-Nitrophenyl)Benzamide

Precaución: for Research Use Only

Formula: C₁₃H₉ClN₂O₃
MDL Number: MFCD01212006
CAS Number: 205827-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR