7-Bromo-5-chloro-1,2-benzoxazole 97%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-5-chlorobenzo[d]isoxazole

A302112-1G A302112-250MG
77154-194EA 284.28 USD
77154-194 77154-196
7-Bromo-5-chloro-1,2-benzoxazole 97%
7-Bromo-5-chloro-1,2-benzoxazole

Precaución: for Research Use Only

Formula: C₇H₃BrClNO
MW: 232.462 g/mol
MDL Number: MFCD11847343
CAS Number: 1260903-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR