2,3-Dibromo-5-chloropyridine 98%

Supplier: Ambeed
Warning

A301733-100G A301733-10G A301733-25G A301733-500G A301733-5G
77154-180EA 105.06 USD
77154-180 77154-182 77154-184 77154-186 77154-188
2,3-Dibromo-5-chloropyridine 98%
2,3-Dibromo-5-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₂Br₂ClN
MW: 271.3400 g/mol
MDL Number: MFCD08687254
CAS Number: 137628-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR