PF-562271 98+%

Supplier: Ambeed
Warning

Synonyms: N-Methyl-N-(3-(((2-((2-oxoindolin-5-yl)amino)-5-(trifluoromethyl)pyrimidin-4-yl)amino)methyl)pyridin-2-yl)methanesulphonamide

A301515-10MG A301515-1MG A301515-2MG A301515-5MG
77154-176EA 210 USD
77154-176 77154-178 77612-734 77612-736
PF-562271 98+%
PF-562271

Precaución: for Research Use Only

Formula: C₂₁H₂₀F₃N₇O₃S
MW: 507.5 g/mol
MDL Number: MFCD16038299
CAS Number: 717907-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR