2-Fluoronicotinonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 3-Cyano-2-fluoropyridine

A301129-100G A301129-10G A301129-1G A301129-25G A301129-5G
77154-168EA 103 USD
77154-168 77154-170 77154-172 77154-174 77685-857
2-Fluoronicotinonitrile 98%
2-Fluoronicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₃FN₂
MW: 122.1 g/mol
MDL Number: MFCD03095082
CAS Number: 3939-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR