PF-04691502 98+%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-8-(rel-(1r,4r)-4-(2-hydroxyethoxy)cyclohexyl)-6-(6-methoxypyridin-3-yl)-4-methylpyrido[2,3-d]pyrimidin-7(8H)-one

A300869-10MG A300869-1MG A300869-50MG A300869-5MG
77154-160EA 103 USD
77154-160 77154-162 77154-164 77154-166
PF-04691502 98+%
PF-04691502

Precaución: for Research Use Only

Formula: C₂₂H₂₇N₅O₄
MW: 425.48 g/mol
MDL Number: MFCD18782794
CAS Number: 1013101-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR