PSI-7977 98%

Supplier: Ambeed
Warning

Synonyms: (S)-Isopropyl 2-(((S)-(((2R,3R,4R,5R)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-4-fluoro-3-hydroxy-4-methyltetrahydrofuran-2-yl)methoxy)(phenoxy)phosphoryl)amino)propanoate, Sofosbuvir

A298621-5G A298621-5MG A298621-250MG A298621-100MG A298621-10MG A298621-50MG A298621-1G A298621-25G
77154-144EA 305 USD
77154-144 77154-146 77612-722 77612-718 77612-720 77685-854 77892-601 77892-602
PSI-7977 98%
PSI-7977

Precaución: for Research Use Only

Formula: C₂₂H₂₉FN₃O₉P
MDL Number: MFCD26523124
CAS Number: 1190307-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR