4-Carbamoyl-piperazine-1-carboxylic acid tert-butyl ester 95%

Supplier: Ambeed
Warning

A297380-250MG A297380-1G
77154-142EA 44.29 USD
77154-142 77685-850
4-Carbamoyl-piperazine-1-carboxylic acid tert-butyl ester 95%
4-Carbamoyl-piperazine-1-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₉N₃O₃
MW: 229.27 g/mol
MDL Number: MFCD06796406
CAS Number: 883554-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR