Midostaurin 99%

Supplier: Ambeed
Warning

Synonyms: PKC412, N-((5S,6R,7R,9R)-6-Methoxy-5-methyl-14-oxo-6,7,8,9,15,16-hexahydro-5H,14H-17-oxa-4b,9a,15-triaza-5,9-methanodibenzo[b,h]cyclonona[jkl]cyclopenta[e]-as-indacen-7-yl)-N-methylbenzamide

A297057-100MG A297057-1MG A297057-1G A297057-250MG
77154-138EA 66 USD
77154-138 77154-140 77593-800 77685-847
Midostaurin 99%
Midostaurin

Precaución: for Research Use Only

Formula: C₃₅H₃₀N₄O₄
MDL Number: MFCD12828879
CAS Number: 120685-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR