4,5-Dichloro-3H-1,2-dithiol-3-one 97%

Supplier: Ambeed
Danger

A296659-25G A296659-5G
77154-126EA 45 USD
77154-126 77154-128
4,5-Dichloro-3H-1,2-dithiol-3-one 97%
4,5-Dichloro-3H-1,2-dithiol-3-one

Precaución: for Research Use Only

Formula: C₃Cl₂OS₂
MDL Number: MFCD00043025
CAS Number: 1192-52-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR