3-Chloro-5-(hydroxymethyl)benzonitrile 98%

Supplier: Ambeed
Warning

A295670-100MG A295670-1G A295670-250MG A295670-5G
77154-116EA 13.39 USD
77154-116 77154-118 77154-120 77154-122
3-Chloro-5-(hydroxymethyl)benzonitrile 98%
3-Chloro-5-(hydroxymethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₆ClNO
MW: 167.592 g/mol
MDL Number: MFCD18392447
CAS Number: 1021871-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR