4,6-Dichloro-5-fluoropyrimidin-2-amine 95%

Supplier: Ambeed
Warning

A295220-100MG A295220-1G A295220-250MG
77154-106EA 6 USD
77154-106 77154-108 77154-110
4,6-Dichloro-5-fluoropyrimidin-2-amine 95%
4,6-Dichloro-5-fluoropyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₄H₂Cl₂FN₃
MDL Number: MFCD11870221
CAS Number: 15598-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR