Methyl 2-[4-(trifluoromethyl)phenyl]acetate 98%

Supplier: Ambeed
Warning

A294047-1G A294047-5G
77154-094EA 25 USD
77154-094 77901-611
Methyl 2-[4-(trifluoromethyl)phenyl]acetate 98%
Methyl 2-[4-(trifluoromethyl)phenyl]acetate

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂
MDL Number: MFCD08669944
CAS Number: 135325-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR