1-Bromo-2-fluoro-4-iodobenzene 98%

Supplier: Ambeed
Warning

A293986-100G A293986-10G A293986-1G A293986-25G A293986-5G
77154-084EA 106 USD
77154-084 77154-086 77154-088 77154-090 77154-092
1-Bromo-2-fluoro-4-iodobenzene 98%
1-Bromo-2-fluoro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrFI
MDL Number: MFCD00042577
CAS Number: 136434-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR