Evacetrapib 98%

Supplier: Ambeed
Danger

Synonyms: 4-trans-(((S)-5-((3,5-Bis(trifluoromethyl)benzyl)(2-methyl-2H-tetrazol-5-yl)amino)-7,9-dimethyl-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)methyl)cyclohexanecarboxylic acid, trans-4-[[(5S)-5-[[[3,5-bis(trifluoromethyl)phenyl]methyl](2-methyl-2H-tetrazol-5-yl)amino]-2,3,4,5-tetrahydro-7,9-dimethyl-1H-1-benzazepin-1-yl]methyl]- cyclohexanecarboxylic acid, LY2484595

A293328-10MG A293328-1MG A293328-5MG
77154-058EA 52.53 USD
77154-058 77154-060 77154-062
Evacetrapib 98%
Evacetrapib

Precaución: for Research Use Only

Formula: C₃₁H₃₆F₆N₆O₂
MW: 638.65 g/mol
MDL Number: MFCD23105886
CAS Number: 1186486-62-3
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR