(R)-Pyrrolidine-3-carbonitrile hydrochloride 98%

Supplier: Ambeed
Danger

A293263-1G A293263-100MG A293263-250MG A293263-5G
77154-056EA 242 USD
77154-056 77892-590 77892-591 77892-592
(R)-Pyrrolidine-3-carbonitrile hydrochloride 98%
(R)-Pyrrolidine-3-carbonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉ClN₂
MDL Number: MFCD11656839
CAS Number: 1153950-54-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR