Calcium chloride dihydrate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Calcium Chloride Dihydrate, Calcium dichloride
A292646-1000G
A292646-500G
77154-048EA
34
USD
77154-048
77154-050
Calcium chloride dihydrate 98%
Calcium chloride dihydrate
Precaución: for Research Use Only
Formula:
CaCl₂·2H₂O MW: 147.01 g/mol Boiling Pt: 1600 Melting Pt: 176 |
MDL Number:
MFCD00149613 CAS Number: 10035-04-8 Merck Index: 13,01660 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...