Quercetin, anhydrous 97%

Supplier: Ambeed
Danger

Synonyms: 3-5-7-3-4-Pentahydroxyflavone, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-1-benzopyran-4-one, 3,5,7,3',4'-Pentahydroxyflavone

A292006-100G A292006-10G A292006-1MG A292006-25G A292006-5G
77154-024EA 37.08 USD
77154-024 77154-026 77154-028 77154-030 77154-032
Quercetin, anhydrous 97%
Quercetin

Precaución: for Research Use Only

Formula: C₁₅H₁₀O₇
MW: 302.25 g/mol
MDL Number: MFCD00006828
CAS Number: 117-39-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR