1-Bromo-4-(phenylethynyl)benzene 97%

Supplier: Ambeed
Warning

A290984-1G A290984-250MG A290984-5G A290984-25G
77153-994EA 13 USD
77153-994 77153-996 77153-998 77901-288
1-Bromo-4-(phenylethynyl)benzene 97%
1-Bromo-4-(phenylethynyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₉Br
MDL Number: MFCD01596910
CAS Number: 13667-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR