2-Chloro-5-methylphenyl isocyanate 98%

Supplier: Ambeed
Danger

A290680-1G A290680-25G A290680-5G
77153-988EA 21.63 USD
77153-988 77153-990 77153-992
2-Chloro-5-methylphenyl isocyanate 98%
2-Chloro-5-methylphenyl isocyanate

Precaución: for Research Use Only

Formula: C₈H₆ClNO
MW: 167.59 g/mol
MDL Number: MFCD01863693
CAS Number: 40398-03-6
UN: 2206
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR