2,5-Dibromobenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 2,5-Dibromobenzene-1-sulphonyl chloride

A290255-25G A290255-5G
77153-972EA 106.09 USD
77153-972 77153-974
2,5-Dibromobenzenesulfonyl chloride 98%
2,5-Dibromobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃Br₂ClO₂S
MW: 334.42 g/mol
MDL Number: MFCD00060685
CAS Number: 23886-64-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR