Methyl 2-(4-(pyrimidin-2-yl)piperazin-1-yl)acetate 97+%

Supplier: Ambeed
Danger

A290025-1G A290025-250MG
77153-968EA 141.11 USD
77153-968 77153-970
Methyl 2-(4-(pyrimidin-2-yl)piperazin-1-yl)acetate 97+%
Methyl 2-(4-(pyrimidin-2-yl)piperazin-1-yl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₄O₂
MW: 236.27 g/mol
MDL Number: MFCD05071221
CAS Number: 896213-29-9
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR