H-Orn(boc)-OH 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Orn(Boc)-OH, N-α-t-Boc-L-Ornithine, Boc-L-Orn-OH, Ndelta-Boc-L-ornithine, (S)-2-Amino-5-((tert-butoxycarbonyl)amino)pentanoic acid
A289711-1G
A289711-250MG
A289711-5G
A289711-100G
A289711-10G
A289711-25G
77153-956EA
6
USD
77153-956
77153-958
77153-960
77901-228
77901-683
77901-980
H-Orn(boc)-OH 95%
H-Orn(boc)-OH
Precaución: for Research Use Only
Formula:
C₁₀H₂₀N₂O₄ |
MDL Number:
MFCD00037219 CAS Number: 13650-49-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...