3-Aminocyclopentanecarboxylic acid 95%

Supplier: Ambeed
Warning

A288129-100MG A288129-250MG A288129-5G A288129-1G
77153-942EA 18 USD
77153-942 77153-944 77153-946 77892-570
3-Aminocyclopentanecarboxylic acid 95%
3-Aminocyclopentanecarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MDL Number: MFCD01318271
CAS Number: 89614-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR