CB5083 99+%

Supplier: Ambeed
Warning

Synonyms: CB-5083, 1-(4-(benzylamino)-7,8-dihydro-5H-pyrano[4,3-d]pyrimidin-2-yl)-2-methyl-1H-indole-4-carboxamide

A287921-10MG A287921-50MG A287921-5MG A287921-1MG A287921-2MG
77153-936EA 109.18 USD
77153-936 77153-938 77153-940 77612-656 77612-658
CB5083 99+%
CB5083

Precaución: for Research Use Only

Formula: C₂₄H₂₃N₅O₂
MW: 413.47 g/mol
MDL Number: MFCD28963914
CAS Number: 1542705-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR