N-Methoxy-N-methyl-cyclobutanecarboxamide 98%

Supplier: Ambeed
Warning

Synonyms: N-Methoxy-N-methylcyclobutanecarboxamide

A287894-250MG A287894-5G A287894-1G
77153-934EA 9.27 USD
77153-934 77593-764 77685-821
N-Methoxy-N-methyl-cyclobutanecarboxamide 98%
N-Methoxy-N-methyl-cyclobutanecarboxamide

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MW: 143.184 g/mol
MDL Number: MFCD12546237
CAS Number: 640768-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR