EW-7197 98%

Supplier: Ambeed
Warning

A287741-2MG A287741-5MG
77153-930EA 12.36 USD
77153-930 77153-932
EW-7197 98%
EW-7197

Precaución: for Research Use Only

Formula: C₂₂H₁₈FN₇
MDL Number: MFCD28348363
CAS Number: 1352608-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR