Methyl 4-amino-2-(trifluoromethyl)benzoate 98%

Supplier: Ambeed
Warning

A287211-250MG A287211-5G
77153-926EA 44 USD
77153-926 77153-928
Methyl 4-amino-2-(trifluoromethyl)benzoate 98%
Methyl 4-amino-2-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₂
MDL Number: MFCD14697988
CAS Number: 894796-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR