tert-Butyl 2-thia-6-azaspiro[3.3]heptane-6-carboxylate 2,2-dioxide 95+%

Supplier: Ambeed
Danger

A287117-100MG A287117-1G A287117-250MG A287117-5G
77153-918EA 74.16 USD
77153-918 77153-920 77153-922 77153-924
tert-Butyl 2-thia-6-azaspiro[3.3]heptane-6-carboxylate 2,2-dioxide 95+%
tert-Butyl 2-thia-6-azaspiro[3.3]heptane-6-carboxylate 2,2-dioxide

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₄S
MW: 247.311 g/mol
MDL Number: MFCD18839239
CAS Number: 1291487-31-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR