Ethyl 1-methyl-1H-pyrrole-2-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A285843-100MG
A285843-1G
A285843-250MG
A285843-5G
77153-908EA
22
USD
77153-908
77153-910
77153-912
77153-914
Ethyl 1-methyl-1H-pyrrole-2-carboxylate 95%
Ethyl 1-methyl-1H-pyrrole-2-carboxylate
Precaución: for Research Use Only
Formula:
C₈H₁₁NO₂ |
MDL Number:
MFCD02179573 CAS Number: 23466-27-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...