3-(Trimethoxysilyl)propyl 2-Bromo-2-methylpropanoate 96%

Supplier: Ambeed
Warning

Synonyms: 3-(Trimethoxysilyl)propyl 2-bromo-2-methylpropanoate, 2-Bromo-2-methylpropanoic acid 3-(Trimethoxysilyl)propyl ester

A284589-100MG A284589-1G A284589-250MG A284589-5G
77153-890EA 13 USD
77153-890 77153-892 77153-894 77153-896
3-(Trimethoxysilyl)propyl 2-Bromo-2-methylpropanoate 96%
3-(Trimethoxysilyl)propyl 2-Bromo-2-methylpropanoate

Precaución: for Research Use Only

Formula: C₁₀H₂₁BrO₅Si
MW: 329.26 g/mol
MDL Number: MFCD09953896
CAS Number: 314021-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR