2',3'-Dichloro-2,2,2-trifluoroacetophenone 95+%

Supplier: Ambeed
Warning

A283764-1G A283764-250MG A283764-5G
77153-882EA 137 USD
77153-882 77153-884 77153-886
2',3'-Dichloro-2,2,2-trifluoroacetophenone 95+%
2',3'-Dichloro-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₃Cl₂F₃O
MDL Number: MFCD02260843
CAS Number: 886371-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR