3,3'-Diethylthiacarbocyanine iodide [DiSC2(3)] 98%

Supplier: Ambeed
Warning

Synonyms: DiSC2(3), 3-Ethyl-2-[3-[3-ethyl-2(3H)-benzothiazolylidene]-1-propenyl]benzothiazolium Iodide, 3,3-Diethylthiacarbocyanine iodide, 3-Ethyl-2-(3-(3-ethylbenzo[d]thiazol-2(3H)-ylidene)prop-1-en-1-yl)benzo[d]thiazol-3-ium iodide

A282616-5G A282616-1G A282616-250MG
77153-864EA 417.15 USD
77153-864 77612-626 77612-628
3,3'-Diethylthiacarbocyanine iodide [DiSC2(3)] 98%
3,3'-Diethylthiacarbocyanine iodide [DiSC2(3)]

Precaución: for Research Use Only

Formula: C₂₁H₂₁IN₂S₂
MW: 492.44 g/mol
MDL Number: MFCD00011956
CAS Number: 905-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR