1,3-Dimethyl-6-amino-1H-indazole 95%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dimethyl-1H-indazol-6-amine

A282549-100MG A282549-250MG A282549-5G
77153-858EA 66.95 USD
77153-858 77153-860 77153-862
1,3-Dimethyl-6-amino-1H-indazole 95%
1,3-Dimethyl-6-amino-1H-indazole

Precaución: for Research Use Only

Formula: C₉H₁₁N₃
MW: 161.21 g/mol
MDL Number: MFCD18432764
CAS Number: 221681-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR