Fmoc-Ser(Ac)-OH 97%

Supplier: Ambeed
Warning

A282200-10G A282200-1G A282200-250MG A282200-25G A282200-5G
77153-844EA 59 USD
77153-844 77153-846 77153-848 77153-850 77685-796
Fmoc-Ser(Ac)-OH 97%
Fmoc-Ser(Ac)-OH

Precaución: for Research Use Only

Formula: C₂₀H₁₉NO₆
MDL Number: MFCD00077070
CAS Number: 171778-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR