Ethyl 8-bromoimidazo[1,2-a]pyridine-2-carboxylate 98%

Supplier: Ambeed
Warning

A281032-100MG A281032-1G A281032-250MG A281032-5G
77153-822EA 10.3 USD
77153-822 77153-824 77153-826 77153-828
Ethyl 8-bromoimidazo[1,2-a]pyridine-2-carboxylate 98%
Ethyl 8-bromoimidazo[1,2-a]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂O₂
MW: 269.09 g/mol
MDL Number: MFCD11976183
CAS Number: 1038393-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR